Learn More
ALADDIN SCIENTIFIC CORPORATION Chitosan, 100g
Supplier: ALADDIN SCIENTIFIC CORPORATION C105799100G
Forms gels with multivalent anions Gives clear solutions that dry to strong clear films Biocompatible antibacterial and environmentally friendly polyelectrolyte with a variety of applications including water treatment chromatography additives for cosmetics textile treatment for antimicrobial activitynovel fibers for textiles photographic papers biodegradable films3 biomedical devices and microcapsule implants for controlled release in drug delivery
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.