Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ascorbic Acid (L-Ascorbic Acid; Vitamin C), Reagents

Catalog No. 01337476
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-476 500 g
01-337-479 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-476 Supplier Reagents Holdings Llc Supplier No. C2144800500C2
Add to Cart
Add to Cart

Ascorbic Acid (L-Ascorbic Acid; Vitamin C), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00064328
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
Synonym Vitamin C, L-Ascorbic Acid
IUPAC Name (5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

Specifications

Boiling Range 190°C
Melting Point 190°C
Color Colorless/White
Boiling Point 190°C
Melting Range 190°C
Packaging Glass Bottle
Quantity 500 g
Specific Gravity 1.65
Grade Reagent
Physical Form Solid
Chemical Name or Material Ascorbic Acid (L-Ascorbic Acid, Vitamin C)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.