Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ascorbic Acid, USP, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18600777 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-777 25 g Amber Glass Bottle
18-600-782 100 g Amber Glass Bottle
18-600-778 500 g Amber Glass Bottle
18-600-779 1 kg Amber Glass Bottle
18-600-780 5 kg Amber Glass Bottle
18-600-781 12 kg Amber Glass Bottle
18-600-783 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-600-777 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. AS10225GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

AS102, 50-81-7, C6H8O6

Spectrum™ Chemical Ascorbic Acid, USP , Vitamin C, is a dietary supplement used to treat and prevent vitamin C deficiency. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00064328
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
IUPAC Name (5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Packaging Amber Glass Bottle
Quantity 25 g
Specific Rotation +20.5° to +21.5°
Percent Purity 99 to 100.5%
Grade USP
Physical Form Crystals or Flakes
Chemical Name or Material Ascorbic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.