Learn More
Description
Specifications
Specifications
| Assay Range | 39.0625 pg/mL |
| Assay Sensitivity | 39.06-2500 pg/mL |
| Assay | Sandwich ELISA |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | Antibody Pairs |
| Specificity | This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human ASGPR1. |
| Target Species | Human |
| For Use With (Application) | Sandwich ELISA |
| Gene Alias | ASGPR, ASGPR 1, ASGP-R 1, asialoglycoprotein receptor 1, CLEC4H1ASGPR1, C-type lectin domain family 4 member H1, Hepatic lectin H1, HL-1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
