Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASGR1 Monoclonal Antibody (8D7), FITC
GREENER_CHOICE

Catalog No. FERMA548985 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548985 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548985 Supplier Invitrogen™ Supplier No. MA548985
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

The monoclonal antibody 8D7 is cross reactive with human ASGPR.

The asialoglycooprotein receptor (ASGPR) is a transmembrane glycoprotein (42 kDa) which mediates binding, internalization and degradation of extracellular glycoproteins that have exposed terminal galactose residues. The receptor is expressed on the surface of the hepatocytes in a polar manner, i. e. it is present on the sinusoidal and lateral plasma membranes but not on the bile canalicular membrane. The mammalian hepatic ASGPR mediate the endocytosis and degradation of serum proteins from which terminal sialic residues have been removed. The rat asialoglycoprotein receptor (ASGPR) consists of three polypeptide subunits: rat hepatic lectin (RHL)-1, RHL-2 and RHL-3. The monoclonal antibody 8D7 recognizes a subunit-specific epitope of RHL-1 of rat ASGPR. The monoclonal antibody 8D7 is cross reactive with human ASGPR.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASGR1
Applications Flow Cytometry, Inhibition Assays, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 8D7
Concentration 100 μg/mL
Conjugate FITC
Formulation PBS with 1% BSA and 0.02% sodium azide
Gene Asgr1
Gene Accession No. P02706
Gene Alias ASGPR; ASGPR 1; ASGP-R 1; ASGPR1; ASGR; ASGR1; Asgr-1; asialoglycoprotein receptor (RHL1); asialoglycoprotein receptor 1; Asialoglycoprotein receptor 1 (hepatic lectin); asialoglycoprotein receptor RHL1; CLEC4H1; C-type lectin domain family 4 member H1; Hepatic lectin 1; hepatic lectin 1 RHL1; hepatic lectin 1, RHL1; hepatic lectin H1; HL-1; RATRHL1; RHL1; RHL-1; Unknown (protein for MGC:133691)
Gene Symbols Asgr1
Host Species Mouse
Immunogen Crude rat liver membrane extracts.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24210
Target Species Rat
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.