Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASGR2 Recombinant Rabbit Monoclonal Antibody (002), PE
GREENER_CHOICE

Catalog No. PIMA540915
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA540915 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA540915 Supplier Invitrogen™ Supplier No. MA540915
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

May be stored for twelve months without detectable loss of activity. Protect from prolonged exposure to light and do not freeze. Sodium azide is toxic to cells and should be disposed of properly so flush with large volumes of water during disposal.

ASGR2 is a cell surface receptor that binds to galactose-terminated glycoproteins. It transports these glycoproteins via a series of membrane vesicles and tubules to an acidic-sorting organelle where the receptor and ligand dissociates. Then the receptor is recycled back to the cell surface. There are four alternatively spliced transcript variants of this gene. This gene has multiple polyadenylation sites.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASGR2
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone 002
Concentration 0.1 mg/mL
Conjugate PE
Formulation PBS with 0.5% BSA and 0.03% ProClin 300
Gene ASGR2
Gene Accession No. P07307
Gene Alias ANCO1; ANCO-2; ASGPR 2; ASGP-R 2; ASGPR2; ASGP-R2; Asgr; Asgr2; Asgr-2; Asialoglycoprotein receptor 2; CLEC4H2; C-type lectin domain family 4 member H2; GAC-1; HBxAg-binding protein; HBXBP; Hepatic lectin 2; hepatic lectin 2 RHL2; hepatic lectin 2, RHL2; hepatic lectin H2; Hepatic lectin R2/3; HL-2; mHL-2; Nbla00144; RHL-2
Gene Symbols ASGR2
Host Species Rabbit
Immunogen Recombinant Human ASGR2/ASGPR2 Protein.
Purification Method Protein A
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 433
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.