Learn More
Restek™ ASI Replacement HPLC Pump Piston Rods
SDP
Replace HPLC Pump Piston Rods to avoid increased system pressure and/or irregular or poor peak shape.
Supplier:  Restek™ 25198
 View more versions of this product
Catalog No. 06-712-754
Only null left
Add to Cart
Description
Description
Inspect piston rod every time piston seals are replaced with a 10x magnifier or microscope to ensure that it does not have wear or scratches and replace it if necessary.
Specifications
Specifications
Model 1090 ASI replacement HPLC pump
Sapphire
Pump Piston Rod
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.