Learn More
AGI NORTH AMERICA INC Syrris Asia Syring Pump provides an ultra-smooth flow rate of 1.0μL to 10mL/min and pressures of up to 20bar (300psi)
SDP
Supplier:  AGI NORTH AMERICA INC 40001AFBF
Encompass
Designed specifically for flow chemistry, the Asia Syringe Pump provides incredibly smooth flow rates. The unique pumping regime enables the continuous pumping of low vapor pressure and low boiling point solvents which are problematic with HPLC-like pumpsThe compact microfluidic module offers two independent flow channels each with an integrated pressure sensor for accurate flow rates.-Wide dynamic flow rate range (1.0uL/min to 10mL/min)-Excellent chemical resistance-Pressure Rating 20bar (300psi)-Easy to replace valve, pressure sensor and syringes without tools-Can be controlled via front panel or PC softwareIncluded:-Asia Syringe Pump with Pump Valves and Pressure Sensors-Asia Red Syringes (2.5mL/5mL) – Flow rate range 50uL to 10mL/minOrder informationThe Asia Syringe Pump can be supplied with alternative syringe sizes for precise and accurate control of lower flow rates.
Catalog No. NC2417596
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.