Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595730 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595730 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595730 Supplier Invitrogen™ Supplier No. PA595730
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Positive Samples: THP-1, HepG2, U-251MG, MCF-7, Mouse brain, Mouse liver, Rat brain, Rat liver; Cellular Location: Lysosome Immunogen sequence: PARLHRIVPR LRDVFGWGNL TCPICKGLFT AINLGLKKEP NVARVGSVAI KLCNLLKIAP PAVCQSIVHL FEDDMVEVWR RSVLSPSEAC GLLLGSTCGH WDIFSSWNIS LPTVPKPPPK PPSPPAPGAP VSRILFLTDL HWDHDYLEGT DPDCADPLCC RRGSGLPPAS RPGAGYWGEY SKCDLPLRTL ESLLSGLGPA GPFDMVYWTG DIPAHDVWHQ TRQDQLRALT TVTALVRKFL GPVPVYPAVG NHESTPVNSF PPPFIEGNHS S.

The protein encoded by this gene is a lysosomal acid sphingomyelinase that converts sphingomyelin to ceramide. The encoded protein also has phospholipase C activity. Defects in this gene are a cause of Niemann-Pick disease type A (NPA) and Niemann-Pick disease type B (NPB). Three transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASM
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene Smpd1
Gene Accession No. P17405, Q04519
Gene Alias Acid sphingomyelinase; ASM; aSMase; A-SMase; NPD; Smpd1; Sphingomyelin phosphodiesterase; sphingomyelin phosphodiesterase 1; sphingomyelin phosphodiesterase 1, acid lysosomal; Zn-SMase
Gene Symbols Smpd1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 70-340 of human SMPD1 / ASM (NP_0005343).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20597, 308909, 6609
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.