Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASNA1 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1B7C11, Proteintech
SDP

Catalog No. 501736777 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6777 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6777 Supplier Proteintech Group Inc Supplier No. 663461IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ASNA1 is the human homolog of the bacterial arsA gene. In E. coli, ArsA ATPase is the catalytic component of a multisubunit oxyanion pump that is responsible for resistance to arsenicals and antimonials.

Specifications

Antigen ASNA1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1B7C11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ASNA1
Gene Accession No. O43681, O54984
Gene Alias ARSA, ARSA I, ARSA1, Arsenical pump driving ATPase, Arsenite stimulated ATPase, ASNA1, ATPase ASNA1, hARSA I, hASNA I, TRC40
Gene Symbols Asna1
Host Species Mouse
Immunogen ASNA1 Fusion Protein Ag7936
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100520362, 288919, 439, 56495
Target Species Human, Mouse, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.