Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASNSD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567372
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567372 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567372 Supplier Invitrogen™ Supplier No. PA567372
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KLRRTRICHLI RPLDTVLDDS IGCAVWFASR GIGWLVAQEG VKSYQSNAKV VLTGIGADEQ LAGYSRHRVR FQSHGLEGLN KE Highest antigen sequence identity to the following orthologs - mouse 84%, rat 82%.

ASNSD1 (Asparagine Synthetase Domain Containing 1) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include asparagine synthase (glutamine-hydrolyzing) activity. An important paralog of this gene is ENSG00000286165. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASNSD1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ASNSD1
Gene Accession No. Q9NWL6
Gene Alias ASNSD1; asparagine synthetase domain containing 1; asparagine synthetase domain-containing protein 1; HCV NS3-transactivated protein 1; NBLA00058; NS3TP1
Gene Symbols ASNSD1
Host Species Rabbit
Immunogen Recombinant Human ASNSD1. Recombinant protein control fragment (Product #RP-108032).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54529
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.