Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ASP 3026
GREENER_CHOICE

Catalog No. p-7110809
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-101-0 10 mg
53-105-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 53-101-0 Supplier Tocris Bioscience™ Supplier No. 5310/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent anaplastic lymphoma kinase (ALK) inhibitor; also potent ACK inhibitor

ASP 3026 is a potent anaplastic lymphoma kinase (ALK) inhibitor (IC50 = 3.5 nM). Also inhibits Ack and ROS1 activity (IC50 values are 5.8 and 8.9 nM respectively). Attenuates proliferation, and induces apoptosis of NPM-ALK+ T cell anaplastic large-cell lymphoma (ALCL) cells in vitro. Suppresses tumor growth of NPM-ALK+ ALCL cell xenografts in mice. Causes tumor shrinkage in hEML4-ALK transgenic mice. Orally available.

Specifications

Chemical Name or Material N2-[2-Methoxy-4-[4-(4-methyl-1-piperazinyl)-1-piperidinyl]phenyl]-N4-[2-[(1-methylethyl)sulfonyl]phenyl]-1,3,5-triazine-2,4-diamine
CAS 1097917-15-1
Quantity 10 mg
Target ALK Inhibitors
Molecular Formula C29H40N8O3S
Formula Weight 580.74
Purity >99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.