Learn More
CPC Scientific H-Asp-Phe-OMe (trifluoroacetate salt) 25MG
SDP
Supplier:  CPC Scientific ASPA001B
Encompass
This dipeptide is 180 to 200 times sweeter than that of sucrose.ONE-LETTER SEQUENCE: DF-OMeMOLECULAR FORMULA: C14H18N2O5MOLECULAR WEIGHT:294.31STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [22839-47-0], RESEARCH AREA: ObesityREFERENCES: Xia Li, et al. Chem Senses, Jun 2011; 36: 453-475.
Catalog No. 50-210-8790
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.