Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Aspartate Aminotransferase Activity Assay Kit, Fluorometric, 1 Kit
Description
- Assay type: Enzyme activity
- Detection method: Fluorescent
- Sensitivity: 2 mU/mL
- Sample types: Serum, other biological fluids
- Applications: Enzyme activity assay
- Cited in: 6 publications
- Compatible equipment: Microplate reader
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Aspartate Aminotransferase |
| For Use With (Equipment) | Microplate Reader |
| Format | Kit |
| No. of Reactions | 100 Tests |
| Detection Method | Fluorescence |
| Sample Type | Serum, Other biological fluids |
| Quantity | 1 Kit |
| For Use With (Application) | Enzyme activity assay |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?