Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASPM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567689
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567689 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567689 Supplier Invitrogen™ Supplier No. PA567689
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ASPM Polyclonal Antibody for Western Blot

This gene is the human ortholog of the Drosophila melanogaster 'abnormal spindle' gene (asp), which is essential for normal mitotic spindle function in embryonic neuroblasts. Studies in mouse also suggest a role of this gene in mitotic spindle regulation, with a preferential role in regulating neurogenesis. Mutations in this gene are associated with microcephaly primary type 5. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASPM
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ASPM
Gene Accession No. Q8IZT6
Gene Alias abnormal spindle microtubule assembly; Abnormal spindle protein homolog; Abnormal spindle-like microcephaly-associated protein; ASP; asp (abnormal spindle) homolog, microcephaly associated; Asp homolog; ASPM; Calmbp1; MCPH5
Gene Symbols ASPM
Host Species Rabbit
Immunogen Synthesized peptide derived from human ASPM.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259266
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.