Learn More
AdipoGen Aspyrone
SDP
Supplier:  AdipoGen BVT0029M001
Encompass_Preferred
Chemical. CAS 17398-00-4. Formula C9H12O4. MW 184.2. Isolated from Aspergillus ochraceus. Antibiotic. Shows nematicidal, insecticidal, antibacterial and antifungal activity. Related to asperlactone and aspinonene.
Catalog No. 50-256-2139
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.