Learn More
Teknova Assay Buffer pH 7.4. 1000mL, Sterile. 8 pack.
SDP
Supplier:  Teknova 2A88908PK
Encompass_Preferred
6.4mM EDTA, 5.1mM EGTA,50.0mM HEPES,149.2mM Sodium Chloride,1.0% Triton X-100,1.0% Bovine Serum Albumin (BSA)
Catalog No. 50-238-7329
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.