Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Associated Bag Biohazard/Infectious Waste Bag Liners
SDP

Catalog No. 19804302 Shop All Associated Bag Company Products
Change view
Click to view available options
Color:
Red
Yellow
Thickness:
1.5 mil
2 mil
4 mil
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Thickness
19-804-302 Yellow 2 mil
19-804-304 Red 4 mil
19-804-300 Red 1.5 mil
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-804-302 Supplier Associated Bag Supplier No. 364100Y
Add to Cart
Add to Cart

Specifications

Color Yellow
Includes Biohazard label
For Use With (Application) Safe for landfill or incineration
Thickness 2 mil
Product Type Contaminated Linens Low Density Gusseted Liner
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.