Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Associated Bag Radioactive Bags
SDP

Catalog No. 191202037 Shop All Associated Bag Company Products
Change view
Click to view available options
Material:
LDPE, LLDPE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-120-2037 LDPE, LLDPE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-120-2037 Supplier Associated Bag Supplier No. 365107Y
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Certifications/Compliance Federal regulation 10-20.203
Color Yellow
Length (Metric) 91.4 cm
Weight (English) 44 lb.
Weight (Metric) 19.9 kg
Width (Metric) 61 cm
For Use With (Application) For safe disposal of radioactive waste
Thickness 3 mm
Material LDPE, LLDPE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.