Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Assorted Ball Set of 12
SDPSCI_ED

Catalog No. S139503 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139503 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139503 Supplier United Scientific™ Supplier No. DBLST12A
Add to Cart
Add to Cart
This item is not returnable. View return policy

A comprehensive sit of hooked, drilled, and solid balls.

Comprehensive set of 12 balls for classroom demonstrations.

Contents:

  • 1 drilled aluminum ball, 25 mm diameter
  • 1 drilled copper ball, 25 mm diameter
  • 1 drilled steel ball, 25 mm diameter
  • 1 drilled wood ball, 25 mm diameter
  • 2 drilled brass balls, 25 mm diameter
  • 2 solid steel balls (one plain and one with suspension lugs)
  • 1 brass ball with hook, 38 mm diameter
  • 1 steel ball with hook, 38 mm diameter
  • 1 copper ball with hook, 38 mm diameter
  • 1 cork ball, 38 mm

All drilled balls feature 3 mm countersunk holes so pendulum cord knots can be recessed.

Packed in a plastic storage box.

Specifications

Grade 5 to 12
Format Set
Languages English
Subject Physics

Warning: Cancer and reproductive harm

Prop 65

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.