Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific Assorted CT-Caps for CapTrack™

Catalog No. 22171732 Shop All Globe Scientific Products
Change view
Click to view available options
Color:
Assorted
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-171-732 Assorted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-171-732 Supplier Globe Scientific Supplier No. CT1CAP

Globe Scientific™ Assorted CT-Caps for CapTrack™ Recapping Archiver provide one with a variety colored caps.

Specifications

Autoclavable Autoclavable
Color Assorted
For Use With (Equipment) CT1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.