Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Assorted Metal Cubes
SCI_ED

Catalog No. S12914 Shop All Eisco Products
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S12914 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S12914 Supplier Eisco™ Supplier No. PH0106
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Set of six Assorted Metal Cubes.

  • Comprising brass, lead, iron, copper, aluminum, zinc.
  • Supplied in plastic box.

Specifications

Grade 7 to 12
Dimensions (L x W) 25 x 77 mm (1 x 3 in.)
Includes 6 Cubes (each comprised of a different metal), Plastic Box
Material Brass, Lead, Iron, Copper, Aluminum, Zinc
Quantity 6 Pk.
Class Size 1 Lab Group
Product Type Metal Cubes
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.