Learn More
Protective Industrial Products Assurance™ 12 Mil Butyl Rubber Glove with Smooth Grip - 14 in.
Shop All Protective Industrial Products Inc. ProductsDescription
- 12 Mil Bromine-Butyl Rubber
- The addition of bromine enhances temperature resistance, offering superior durability
- 14 in. length for extended forearm protection
- Smooth Grip
- Protects against hazardous chemicals and materials
- Easy to decontaminate
- Excellent holdout
- Maintains flexibility from -40°C to +90°C (-40°F to +194°F)
- ESD safe 10^8 Ohms
Specifications
Specifications
| Applications | Laboratory |
| Color | Black |
| Latex Free | Yes |
| Material | Butyl |
| Product Type | Butyl Gloves |
| Size | Small |
| Thickness (English) | 12 mil |
| Thickness (Metric) | 0.3048 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.