Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protective Industrial Products Assurance™ 20 Mil Butyl Rubber Glove with Smooth Grip - 14 in.

Catalog No. 18813336 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-813-336 Small
18-813-337 Medium
18-813-338 Large
18-813-339 X-Large
18-813-340 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-813-336 Supplier Protective Industrial Products Supplier No. BUT05/7
Add to Cart
Add to Cart
Request a Quote

Designed to handle extreme exposure to polar hydrocarbons such as ketones, acids, esters, and amine derivatives.

  • 20 Mil Bromine butyl rubber
  • The addition of bromine enhances temperature resistance, offering superior durability
  • 14 in. length for extended forearm protection
  • Smooth grip
  • Protects against hazardous chemicals and materials
  • Easy to decontaminate
  • Excellent holdout
  • Maintains flexibility from -40°C to +90°C (-40°F to +194°F)
  • ESD safe 10^8 Ohms

Specifications

Applications Laboratory
Color Black
Latex Free Yes
Material Butyl
Product Type Butyl Gloves
Size Small
Thickness (English) 20 mil
Thickness (Metric) 0.508 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.