Learn More
Description
- Light weight construction and hand specific design provide real comfort with protection
- Provides flexibility, tactile sensitivity and dexterity sometimes lacking in heavier gauge unsupported nitriles
- Provides the protection from chemical and physical hazards not always found in thinner gauge nitrile disposables
- Unlined to reduce potential of product contamination
- Straight cuff adds additional length to protect the forearm from liquid run-off
- Raised Diamond pattern for an enhanced grip
- Chlorinated to harden and cleanse the surface of the glove
Specifications
Specifications
| Certifications/Compliance | ISO 9001:2000 |
| Chlorinated | No |
| Color | Green |
| Material | Nitrile |
| Size | Medium |
| Thickness (English) | 11 mil |
| Thickness (Metric) | 0.27 mm |
| For Use With (Application) | light chemical applications, printing, meat and poultry processing |
| Glove Cuff Style | Straight |
| Length (English) | 13 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.