Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Assurance™ Unlined Nitrile Gloves

Catalog No. 19121187D Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-121-187D X-Large
19-121-187B Medium
19-121-187C Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-121-187D Supplier PIP™ Supplier No. 50N115G/XL

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in light chemical applications, printing, meat and poultry processing.

  • Light weight construction and hand specific design provide real comfort with protection
  • Provides flexibility, tactile sensitivity and dexterity sometimes lacking in heavier gauge unsupported nitriles
  • Provides the protection from chemical and physical hazards not always found in thinner gauge nitrile disposables
  • Unlined to reduce potential of product contamination
  • Straight cuff adds additional length to protect the forearm from liquid run-off
  • Raised Diamond pattern for an enhanced grip
  • Chlorinated to harden and cleanse the surface of the glove

Specifications

Certifications/Compliance ISO 9001:2000
Chlorinated No
Color Green
Material Nitrile
Size X-Large
Thickness (English) 11 mil
Thickness (Metric) 0.27 mm
For Use With (Application) light chemical applications, printing, meat and poultry processing
Glove Cuff Style Straight
Length (English) 13 in.
Length (Metric) 33 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.