Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Assurance™ Unsupported 21mil Medium and Heavy Weight Nitrile Gloves, Flock Lined

Catalog No. 19162736 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-162-736 Medium
19-161-843 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-162-736 Supplier PIP™ Supplier No. 50N2420GY/M

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provides resistance to physical hazards such as cut, puncture, snag and abrasion and many chemical hazards including solvents and oil-based chemicals

  • Thickness: 21mil
  • Flock lined
  • Raised diamond grip
  • Straight cuff
  • Color: gold over yellow
  • Length12 in. (30.48cm)
  • It is strongly advised not to wash and re-use these gloves when they are used to protect against chemical products.

    • Chemical processing
    • Oil refining and petrochemicals
    • Food processing
    • Aerospace and automotive degreasing
    • Automotive assembly and painting
    • Machining operations
    • Metal fabrication
    • Graphic arts
    • Printing cleanup
    • Furniture manufacturing
    • Electronics

Certifications

ISO 9001:2000

Compliance

Federal regulations

Specifications

Certifications/Compliance ISO 9001:2000, FDA 21 CFR 170-199
Color Gold, Yellow
Material Nitrile
Powdered Powdered
Size Medium
Thickness (English) 21 mil
For Use With (Application) Chemical processing, Oil refining and Petrochemicals, food processing, aerospace and automotive degreasing, automotive assembly and painting, machining operations, Metal fabrication graphic arts and printing cleanup, furniture manufacturing and
Glove Cuff Style Straight
Length (English) 12 in.
Length (Metric) 30.5 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.