Learn More
Getinge USA AccuFast BI Indicators (50/box)
Supplier:  Getinge USA 61301606636
Encompass_Preferred
The Biological Indicator consists of a plastic vial with an inoculated spore strip and sealed glass vial containing media placed inside. The outer vial is sealed with a plastic cap and filter paper. Small orifices in each cap provide for steam penetration. When the glass vial is crushed, the growth media flows onto the inoculated spore strip. The vial is then incubatedand, if viable spores are present, the media color will change color. After exposure to steam, the biological indicator when incubated will either indicate growth or no growth.
Catalog No. NC0757874