Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ AssureTouch™ Agile™ Nitrile Medical Examination Gloves with Textured Fingertips

Catalog No. 17101086 Shop All Medicom Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-101-086 Large
17-101-083 X-Small
17-101-084 Small
17-101-085 Medium
17-101-087 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-101-086 Supplier Medicom™ Supplier No. 9998D
Add to Cart
Add to Cart
This item is not returnable. View return policy

AssureTouch™ Agile™ Nitrile Medical Examination Gloves with Textured Fingertips. This nitrile glove offers optimal puncture and tear resistance without compromising dexterity or tactility. Textured fingertips ensure a secure grip, even when wet. Chemo tested.

  • These nitrile gloves offer superior puncture, tear and chemical resistance without compromising dexterity or tactility.
  • Textured fingertips ensure a secure grip, even when wet.
  • Chemo tested.

Specifications

Chemotherapeutic Rated Yes
Chlorinated No
Accelerator Free No
Applications Healthcare, Industrial, Laboratory
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Fingertip Textured
Finger Thickness (English) 4.5 mil
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Large
Sterility Non-sterile
Thickness (English) 4.5 mil
FDA 510 (k) Yes
Glove Cuff Style Beaded
Model 9998-D
Quantity 1000/Cs.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.