Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ AssureTouch™ Shield Vinyl Medical Examination Gloves

Catalog No. 17000809 Shop All Medicom Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-000-809 Small
17-000-810 Medium
17-000-812 X-Large
17-000-811 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 17-000-809 Supplier Medicom™ Supplier No. 9994A
Add to Cart
Add to Cart
This item is not returnable. View return policy

High-grade vinyl ensures reliable protection. AssureTouch™ Shield Vinyl Medical Examination Gloves provide a silky feel and smooth texture, as well as enhanced tactile sensitivity.

  • High-grade vinyl ensures reliable protection.
  • Silky feel and smooth texture provide enhanced tactile sensitivity.High-grade vinyl ensures reliable protection. Silky feel and smooth texture provide enhanced tactile sensitivity.
  • Clear color

Specifications

Certifications/Compliance FDA510k, ASTM D412, ASTM D5151, ASTM D6124, ASTM D5250, ASTM F1671, ASTM F739
Chlorinated No
Accelerator Free No
Applications Industrial, Laboratory, Healthcare
AQL 1.5
Color Clear
Disposable Single-use
Exterior Finish No Texture
Latex Free Yes
Material Vinyl
Palm Thickness (English) 3.2 mil
Powdered Powder Free
Product Type Vinyl Examination Gloves
Size Small
Sterility Non-sterile
Thickness (English) 3.2 mil
FDA 510 (k) Yes
Glove Cuff Style Beaded
Model 9994A
Series AssureTouch™ Shield™
Product Line AssureTouch Shield
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.