Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ AssureTouch™ Ultra Nitrile Medical Examination Gloves

Catalog No. 17986195 Shop All Medicom Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-986-195 X-Small
17-986-196 Small
17-986-197 Medium
17-986-198 Large
17-986-199 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-986-195 Supplier Medicom™ Supplier No. 9940A
Add to Cart
Add to Cart

Non-sterile, single-use, powder-free, latex-free gloves with textured fingertips, ambidextrous, preventing cross contamination.

  • Provides reliable protection against contamination and exposure to harmful substances in healthcare and laboratory settings.
  • Durable nitrile material offers puncture and chemical resistance while maintaining tactile sensitivity.
  • Compliant with ASTM D6319 and ASTM D6978 standards for quality and safety.
  • Ideal for handling chemotherapy drugs and other critical applications requiring powder-free, latex-free gloves.
  • Textured fingertips enhance grip for precise handling and ambidextrous use in clinical procedures.

Specifications

Chlorinated Yes
Accelerator Free No
Applications Healthcare, Laboratory, Industrial
AQL 2.5
Color Violet, Blue
Disposable Single-use
Exterior Finish Fingertip Textured
Finger Thickness (English) 3.1 mil
Finger Thickness (Metric) 0.08 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 2.3 mil
Palm Thickness (Metric) 0.06 mm
Powdered Powder Free
Product Type Nitrile Examination Glove
Size X-Small
Sterility Non-sterile
Thickness (English) 2.3 mil
Thickness (Metric) 0.06 mm
FDA 510 (k) Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.