Learn More
Apexbio Technology LLC AGI-6780 1432660-47-3 100mg
SDP
Supplier:  Apexbio Technology LLC A3146100
Encompass_Preferred
AGI-6780 (CAS 1432660-47-3) is a potent and selective inhibitor of the oncogenic IDH2/R140Q mutant enzyme exhibiting an IC50 of 23 nM In cellular assays AGI-6780 effectively reduces intracellular 2-hydroxyglutarate (2HG) levels in TF1/R140Q human glioblastoma cells The compound also reverses the differentiation block mediated by IDH2/R140Q in TF1 cells in the presence of erythropoietin restoring erythroid marker expression such as KLF1 and Hemoglobin G1/2 AGI-6780 serves as a valuable tool for studying mutant IDH2-driven metabolic alterations and cellular differentiation processes in cancer research
Catalog No. 50-000-95256
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.