Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Astaxanthin, Antioxidant Agent, 596.8 Da, 5 mg

Catalog No. 502706003
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6003 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6003 Supplier Abcam Supplier No. AB1441935MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent antioxidant agent that acts as a PPARα agonist, PPARγ antagonist, and free radical scavenger.

  • Molecular Weight: 596.8 Da
  • Purity: >97%
  • Potent antioxidant agent
  • PPARα agonist and PPARγ antagonist
  • Biological activity: Free radical scavenger; reduces oxidative stress; induces apoptosis; inhibits NF-κB
  • In vivo effects: Anti-inflammatory, antihypertensive, cardioprotective, neuroprotective and antitumor effects
  • Other: Orally active

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Target Molecule Astaxanthin
Product Type Antioxidant
Content And Storage Store at -20°C.
Formulation 100% Astaxanthin
Quantity 5 mg
For Use With (Application) Antioxidant research, PPARα agonist, PPARγ antagonist, Free radical scavenging, Apoptosis induction, NF-κB inhibition, Inflammation, Hypertension, Cardioprotection, Neuroprotection, Antitumor studies
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.