Learn More
Abcam Astaxanthin, Antioxidant Agent, >97% Purity, 25 mg
Description
- Molecular Weight: 596.8 Da
- Purity: >97%
- Potent antioxidant agent: PPARα agonist and PPARγ antagonist
- Biological activity: Free radical scavenger; reduces oxidative stress; induces apoptosis; inhibits NF-κB
- Effects: Shows anti-inflammatory, antihypertensive, cardioprotective, neuroprotective and antitumor effects in vivo
- Administration: Orally active
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | Astaxanthin |
| Format | Vial |
| Product Type | Antioxidant |
| Buffer | 100% Astaxanthin |
| Content And Storage | Store at -20°C. |
| Formulation | 100% Astaxanthin |
| Quantity | 25 mg |
| For Use With (Application) | Antioxidant studies, PPAR receptor research, Apoptosis research, Inflammation research, Cardiovascular research, Neuroprotection research, Oncology research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.