Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Astec™ CHIROBIOTIC™ T Chiral HPLC Guard Column Cartridge, 5 μm Particle Size

Catalog No. 111009723 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter: 4 mm; Length: 2 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
11-100-9723 Diameter: 4 mm; Length: 2 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-9723 Supplier MilliporeSigma™ Supelco™ Supplier No. 12100AST
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHIROBIOTIC™ T columns utilize teicoplanin as a chiral selector.

CHIROBIOTIC™ T columns offer unique selectivity for a number of classes of molecules, specifically underivatized alpha, beta, gamma and cyclic amino acids, N-derivatized amino acids, hydroxy-carboxylic acids, acidic compounds including carboxylic acids and phenols, small peptides, neutral aromatic analytes and cyclic aromatic and aliphatic amines. Separations normally obtained on a chiral crown ether or ligand exchange-type CSPs are also possible on the CHIROBIOTIC™ T and T2, but in much simpler mobile phases, like water-alcohol. In addition, all of the known beta-blockers (amino alcohols), and dihydrocoumarins have been resolved. CHIROBIOTIC™ T and T2 differ in their bonding chemistry and the pore size of the support particle, giving them different selectivity and preparative capacity.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1/Pk.
Particle Size 5 μm
Length (Metric) 2 cm
Pore Size 100 Å
Type Chiral HPLC Guard Column
Diameter (Metric) Inner 4 mm
Phase Chiral
For Use With (Application) HPLC
Product Line Astec CHIROBIOTIC

Astec and CHIROBIOTIC are registered trademarks of Sigma-Aldrich Co. LLC.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.