Learn More
Enzo Life Sciences Astemizole (10mg). CAS: 68844-77-9
SDP
Supplier:  Enzo Life Sciences BMLNH1010010
Encompass_Preferred
Selective histamine H1 receptor antagonist. Purity: ≥99% (TLC). Solubility: Soluble in chloroform (clear faint yellow solution at 100mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-1192
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.