Learn More
ASTM™ D6584 Standard Solution Kit with Internal Standards, Certified Reference Material, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Quantity | 1 Kit |
| Grade | TraceCERT™ Analytical Standard |
| Concentration or Composition (by Analyte or Components) | ASTM(R) D6584 Standard solution 1, certified reference material,In pyridine (varied conc.); ASTM(R) D6584 Standard Solution 2,In pyridine (varied), ampule of 1 mL; ASTM(R) D6584 Standard Solution 3, certified reference material,In pyridine (varied), ampule of 1 mL; ASTM(R) D6584 Standard Solution 4, certified reference material,In pyridine (varied conc.); ASTM(R) D6584 Standard Solution 5, certified reference material,In pyridine (varied); ASTM(R) D6584 1,2,4-Butanetriol Solution, certified reference material, 1000 μg/mLIn pyridine, ampule of 5 mL; ASTM(R) D6584 Tricaprin Solution, certified reference material, 8000 μg/mLIn pyridine |
ASTM is a registered trademark of American Society for Testing and Materials