Learn More
AdipoGen Astragaloside IV (50 mg)
SDP
Supplier:  AdipoGen CDXA0577M050
Encompass_Preferred
Chemical. CAS: 84687-43-4. Formula: C41H68O14. MW: 784.98. Astragaloside IV (AS-IV) is a bioactive saponin and constituent of the traditional chinese medicine plant Astragali radix. AS-IV has multiple pharmacologic effects, including anti-inflammatory, antioxidative, anti-asthma, antidiabetes, anticancer, immunoregulation, neuroprotective and cardioprotective properties via numerous signaling pathways. This includes inhibition of NF-kappaB, inducing hypoxia-inducible factor-1alpha accumulation via PI3K/Akt pathway, reducing Abeta production in Alzheimer's disease through inhibition of BACE1, working as a natural PPARgamma agonist, improving lipid metabolism in obese mice, enhancing chemosensitivity through B7-H3 inhibition, and suppressing glucose-induced NLRP3 inflammasome activation.
Catalog No. 50-205-4284
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.