Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Astrin/MAP126 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501560580
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-0580 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-0580 Supplier Bethyl Laboratories, Inc Supplier No. A301511AT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

A novel gene product, hMAP126, interacts with p29 in the yeast two-hybrid assay. The subcellular distribution of hMAP126 was localized to the mitotic spindle and is phosphorylated by p34(cdc2) kinase. Human MAP126 is likely involved in the functional and dynamic regulation of mitotic spindles.

Specifications

Antigen Astrin/MAP126
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene SPAG5
Gene Accession No. Q96R06
Gene Alias DEEPEST, MAP126, hMAP126
Gene Symbols SPAG5
Host Species Rabbit
Immunogen Between 1 and 50
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10615
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.