Learn More
Sciencell Research Laboratories Astrocyte Medium, Complete Kit (includes supplements)
Supplier:  Sciencell Research Laboratories 1801
Encompass
Astrocyte medium is a complete medium designed for optimal growth of normal human astrocytes in vitro. It is shipped as a kit with FBS, astrocyte growth supplement, and Pen/Strep solution.
Catalog No. NC9707094