Learn More
Selleck Chemical LLC ASTX-029 S0872-5MG
SDP
Supplier:  Selleck Chemical LLC S08725MG
Encompass_Preferred
ASTX-029 is an orally bioavailable inhibitor of the extracellular signal-regulated kinases (ERK) 1 and 2 with potential antineoplastic activity ASTX-029 inhibits ERK-dependent tumor cell proliferation and survival
Catalog No. 50-217-3052
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.