Learn More
Selleck Chemical LLC Tyrphostin AG 879 S2816-25mg
Supplier: Selleck Chemical LLC S281625MG
Tyrphostin AG 879 potently inhibits HER2/ErbB2 with IC50 of 1 M 100- and 500-fold higher selective to ErbB2 than PDGFR and EGFR
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.