Learn More
Apexbio Technology LLC AT-406 (SM-406), 1071992-99-8, MFCD22124467, 5 mg
SDP
Supplier:  Apexbio Technology LLC A30195
Encompass_Preferred
MF: C32H43N5O4, MW: 561.71, Solubility: 27.65mg/mL in DMSO. AT-406 is alsonamed as SM-406. AT-406 is a potent and orally bioavailable antagonist of multiple inhibitor of apoptosis proteins (IAPs). The inhibitor of apoptosis proteins(IAPs), originally identified in baculoviruses,are a family of apoptosis suppressor proteins which could bind and inhibit specific enzymes (caspases) such as caspase 3, 7, and 9, but not caspase 8. Growing evidence has showed that IAPs can regulateprogrammed cell death, apoptosis, cell division, cell cycle progression, and signal transduction pathways.
Catalog No. 50-115-0463
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.