Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Worldwide™ Protective Products ATA™ Cut Resistant Sleeve 2-ply Design

Catalog No. 18701032 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Standard
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-701-032 Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-701-032 Supplier Worldwide™ Protective Products Supplier No. MSATACM2X2.518T
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sta-Cool™ inner layer wicks moisture away from the arm, keeping the wearer dry, cool and comfortable. Keep the wrist protected.

  • Serious Arm Protection, All Day Comfort
  • Keep arms cool, comfortable and well protected with this ANSI A4 cut resistant sleeve
  • Durability, strong cut and abrasion resistance are provided by ATA™ fiber technology
  • Sta-Cool™ inner layer wicks moisture away from the arm, keeping the wearer dry, cool and comfortable all day
  • Outstanding stretch means this sleeve will conform to just about any size arm – big or small – and can be worn comfortably over clothing
  • Machine washable
  • Made in the USA

Specifications

Size Standard
Product Type Cut-resistant Sleeve
Construction Seamless Ribbed Knit
Material ATA™ Engineered Fiber
Color Yellow
Sterility Non-sterile
Style Ambidextrous
Disposable Reusable
Closure Type Elastic Ends
Seam Type Seamless
Cut Resistance Rating ANSI A4
Latex Free Yes
For Use With (Application) Medium-to-High Risk Hazards
Includes Thumb hole
Length (English) 18 in.
Length (Metric) 45.7 cm
Width (English) 3 in.
Width (Metric) 7.6 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.