Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Worldwide™ Protective Products ATA™/Aramid Fiber Cut and Heat Resistant Gloves

Catalog No. 18701028 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-701-028 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-701-028 Supplier Worldwide™ Protective Products Supplier No. M2KVBK4L
Add to Cart
Add to Cart
This item is not returnable. View return policy

ATA™ Technology utilizes Aramid Fibers in combination with a cotton liner to deliver cut resistance, and intermittent conductive heat protection up to 608°F.

  • Provides protection against heat and cut hazards such as glass, metal, and other parts or equipment
  • 7 gauge outer glove combines ATA™ Technology with Aramid Fibers
  • Soft cotton liner efficiently traps air between the layers for insulation and protection
  • Capable of enduring intermittent conductive heat up to 608°F
  • Made in the USA

Note:There is a minimum order quantity of 1 case.

Specifications

Accelerator Free Yes
Applications Industrial
Coating Material Uncoated
Color Yellow
Cut Resistance Rating ANSI A5
Latex Free Yes
Material ATA Engineered Fiber, Aramid Fiber
Product Type Heat-resistant Gloves
Size Large
Style Standard
Temperature (English) Max. 608°F (Intermittent)
Temperature (Metric) Max 320°C (Intermittent)
For Use With (Application) Food Processing, Metal/Plastics Fabrication, Glass Production, and Oven Work
Glove Cuff Style Continuous Knit
Liner Material Cotton
Series M2KVBK-4
Yarn Gauge 7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.