Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Worldwide™ Protective Products ATA™ M530 Cut-Resistant, High Dexterity Gloves

Catalog No. 18701015 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Glove Cuff Color:
Black
Blue
Brown
Green
Navy
Orange
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Glove Cuff Color
18-701-015 2X-Large Navy
18-701-010 X-Small Orange
18-701-011 Small Blue
18-701-012 Medium Green
18-701-013 Large Brown
18-701-014 X-Large Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 18-701-015 Supplier Worldwide™ Protective Products Supplier No. M530XXL
Add to Cart
Add to Cart
This item is not returnable. View return policy

ANSI A4 cut protection gloves are ideal for picking up broken glass, dissecting samples and working with task-based knives and scalpels. Thin, form-fitting gloves provide the dexterity to precisely handle instruments and equipment.

  • Uses ATA™ fiber technology to deliver a thin, form-fitting and lightweight design that supplies ANSI A4 cut protection appropriate for medium-to-high cut hazards
  • The 13-gauge seamless knit gloves provide the dexterity and fingertip sensitivity you need to pick up broken glass, dissect samples, and work with task-based knives and scalpels
  • Wear them under unlined nitrile gloves to add a cut resistant barrier
  • The ATA™ fiber blend also uses Lycra fibers for lots of stretch, making gloves comfortable enough for extended periods of wear
  • The Hide-Away technology provides long-lasting abrasion resistance
  • Color coded hems make it easy to choose the right glove size

Specifications

Coating Material Uncoated
Color Green
Cut Resistance Rating ANSI A4
Hand Specific
Latex Free Yes
Material ATA Engineered Fiber
Product Type Cut-resistant Gloves
Size 2X-Large
For Use With (Application) Glass Handling, Metal Fabrication, Plastics and Molding, Use as a Liner Glove
Glove Cuff Color Navy
Glove Cuff Style Continuous Cuff
Model M530-S
Series M530
Yarn Gauge 13
Length (English) 10.5 in.
Length (Metric) 26.7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.