Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Worldwide™ Protective Products Foam Nitrile-Coated Heat Producing ATA™ PioNIR™ Fiber Gloves

Catalog No. p-7161861 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-701-020 Small
18-701-019 Medium
18-701-018 Large
18-701-017 X-Large
18-701-016 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 18-701-020 Supplier Worldwide™ Protective Products Supplier No. PIONIRATA13NDS

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Exclusive PioNIR™ Fiber Technology warms gloves without heat packs or batteries. When exposed to sun and certain indoor light, the glove fibers absorb near infrared rays (NIR) and convert them into heat, increasing the glove's temperature by up to 18°F.

  • ATA™ Advanced Technology Armor delivers a high degree of dexterity
  • Foam nitrile coating enhances grip capabilities in wet or dry conditions
  • PIONIR™ fiber technology warms the gloves without heat packs or batteries
  • When exposed to sun and certain indoor light, the glove fiber absorbs near infrared rays (NIR) and converts them into heat
  • Glove temperature can increase by as much as 18°F
  • Machine washable without affecting heat producing technology
  • Ideal for working outdoors in cold conditions

Specifications

ANSI Abrasion Rating ANSI 2
Coating Material Nitrile Foam
Color Gray
Cut Resistance Rating ANSI A2
Hand Specific
Material ATA Engineered Fiber
Size Small
For Use With (Application) Agriculture, Construction, Cold Storage, Food Packaging, and Utilities
Series PIONIR-ATA13ND
Yarn Gauge 13

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.