Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATAD5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140323
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140323 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140323 Supplier Invitrogen™ Supplier No. PA5140323
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: MLLENNKGIK NSFEQKQITQ TKSTNATNSN VKDVGAEEPS RKNATSLILF EEVDVIFDED AGFLNAIKTF MATTKRPVIL TTSDPTFSLM FDGCFEE Highest antigen sequence indentity to the following orthologs: Mouse - 81%, Rat - 83%.

Involved in DNA damage response. Involved in a RAD9A-related damage checkpoint, a pathway that is important in determining whether DNA damage is compatible with cell survival or whether it requires cell elimination by apoptosis. Modulates the RAD9A interaction with BCL2 and thereby induces DNA damages-induced apoptosis. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATAD5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATAD5
Gene Accession No. Q96QE3
Gene Alias ATAD5; ATPase family AAA domain-containing protein 5; ATPase family, AAA domain containing 5; C130052G03Rik; C17orf41; chromosome fragility associated gene 1; chromosome fragility-associated gene 1 protein; ELG1; enhanced level of genomic instability 1 homolog; FLJ12735; FRAG1; Gm17; RGD1561464
Gene Symbols ATAD5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ATAD5. Recombinant protein control fragment (Product #RP-109115).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79915
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.