Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ataxin 3 Monoclonal Antibody (2SCA-1H9)
GREENER_CHOICE

Catalog No. PIMA3082 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA3082 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA3082 Supplier Invitrogen™ Supplier No. MA3082
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA3-082 detects Ataxin 3 from human, mouse and rat samples. MA3-082 has been successfully used in Western blot, immunofluorescence, and ELISA applications. MA3-082 has been successfully used in Western blot, immunofluorescence, and ELISA applications. The MA3-082 immunogen is the human ataxin-3 fragment from aa 112 - aa L249 as a fusion protein.

Machado-Joseph disease is an autosomal dominant neurologic disorder, and is now known to be the same as previously described spinocerebellar ataxia-3. MJD protein (Ataxin-3) contains (CAG)n repeats in the coding region, and the expansion of these repeats from the normal 13-36 to 68-79 is the cause of Machado-Joseph disease. There is a negative correlation between the age of onset and CAG repeat numbers. This protein interacts with key regulators (CBP, p300 and PCAF) of transcription and represses transcription, and also acts as a histone-binding protein that regulates transcription. MJD is a deubiquitinating enzyme.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ataxin 3
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2SCA-1H9
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.05% sodium azide
Gene ATXN3
Gene Accession No. O35815, P54252, Q9CVD2
Gene Alias 2210008M02Rik; AI463012; AI647473; AT3; ataxin 3; ataxin 3 variant an; ataxin 3 variant ao; ataxin 3 variant at; ataxin 3 variant e; ataxin 3 variant h; ataxin 3 variant m; ataxin 3 variant r; ataxin 3 variant ref; ataxin 3 variant y; ataxin3; ataxin-3; ATX3; ATXN3; JOS; josephin; Machado-Joseph disease (spinocerebellar ataxia 3, olivopontocerebellar ataxia 3, autosomal dominant, ataxin 3); Machado-Joseph disease (spinocerebellar ataxia 3, olivopontocerebellar ataxia 3, autosomal dominant, ataxin 3) homolog; Machado-Joseph disease protein 1; Machado-Joseph disease protein 1 homolog; MJD; MJD1; olivopontocerebellar ataxia 3; Rsca3; Sca3; spin; Spinocerebellar ataxia type 3 protein
Gene Symbols ATXN3
Host Species Mouse
Immunogen Human Ataxin-3 fragment from aa F112-aa L249.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110616, 4287, 60331
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.