Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ataxin 7 Monoclonal Antibody (3SCA-1C1)
GREENER_CHOICE

Catalog No. PIMA3083 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA3083 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA3083 Supplier Invitrogen™ Supplier No. MA3083
  • $533.54
    $423.65 / Each of 1

    Save $109.89

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA3-083 detects Ataxin 7 from human and mouse samples. MA3-083 has been successfully used in Western blot, immunofluorescence, and ELISA applications. The MA3-083 epitope is mapped between aa 66 and aa 119.

Spinocerebellar ataxia type 7 (SCA7) belongs to a class of hereditary neurodegenrative diseases characterized by abnormally expanded polyglutamine (CAG) repeats at the N-terminus. This autosomal dominant cerebellar ataxia primarily affects the cerebellum, retina, and brain stem and causes dementia, macular degeneration and other neurodegenerative characteristics. SCA7 gene encodes the ubiquitous 892 residue protein ataxin-7. This protein is typically located in the cytoplasm and on the nuclear membrane of normal brain neurons. In cells where there is a mutation of the SCA7 gene, ataxin 7 accumulates in intranuclear inclusions and can result in cell death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ataxin 7
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3SCA-1C1
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.05% sodium azide
Gene ATXN7
Gene Accession No. O15265, Q8R4I1
Gene Alias A430107N12Rik; ADCAII; AI627028; Ataxin 7; Ataxin7; Ataxin-7; ATXN7; Atxn7-PA; Atxn7-PB; Atxn7-PC; Atxn7-PD; CG9866; CG9866-PA; CG9866-PB; CG9866-PC; CG9866-PD; Dmel\CG9866; Dmel_CG9866; LD40170p; OPCA3; RGD1562692; SCA7; spinocerebellar ataxia 7 homolog; spinocerebellar ataxia type 7 protein; Spinocerebellar ataxia type 7 protein homolog
Gene Symbols ATXN7
Host Species Mouse
Immunogen Recombinant human Ataxin 7 aa 1 - 229.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246103, 6314
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.